missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iododeoxyuridine/IdU Antibody (32D8.D9), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP2-44056-0.05MG
This item is not returnable.
View return policy
Description
Iododeoxyuridine/IdU Monoclonal specifically detects Iododeoxyuridine/IdU in Human, Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Iododeoxyuridine/IdU | |
| Monoclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% sodium azide | |
| Anti-IdU monoclonal antibody was produced in mice by repeated immunizations prepared via immunizations with BromodeoxyUridine-KLH followed by hybridoma development. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Dot Blot | |
| 32D8.D9 | |
| Western Blot 1:2000 - 1:5000, Flow Cytometry 1:10-1:1000, ELISA 1:1000-1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Dot Blot 1:100-1:2000 | |
| Mouse | |
| 0.05 mg | |
| Non-species specific | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction