Learn More
Lenz Laborglasinstrumente™ Iodine Determination Flasks with Conical Ground Joint, without Collar
Made from borosilicate glass. Lenz Laborglasinstrumente™ Iodine Determination Flasks with Conical Ground Joint, without Collar is perfect for adding precise chemical or liquid that needs to go into a solution. You can combine the required reagents in precise quantities to make a solution.
17.45 € - 222.60 €
Specifications
Material | Borosilicate Glass |
---|---|
Includes | Conical Ground Glass Joint |
Shape | Conical |
Standard Taper Joint | NS 29/32 |
Graduated | Yes |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10599961
|
Lenz Laborglasinstrumente™
03026058 |
500 mL |
222.60 €
Pack of 10 |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
10080281
|
Lenz Laborglasinstrumente™
03026049 |
250 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- With hollow hexagonal stopper NS 29/32
- With scale
- With inscription label
- Material: DURAN™
Specifications
Borosilicate Glass | |
Conical | |
Yes |
Conical Ground Glass Joint | |
NS 29/32 |