missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript™ Plus Direct cDNA Labeling System with Alexa Fluor™-aha-dUTPs

Product Code. 10226952
Change view
Click to view available options
Quantity:
30 Reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10226952 30 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10226952 Supplier Invitrogen™ Supplier No. L101506

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

All SuperScript™ Direct cDNA Labeling Systems include SuperScript™ III Reverse Transcriptase, anchored Oligo(dT), random primers, control RNA, 5X first-strand buffer, 0.1M DTT, 10mM dNTP mix, low-elution purification columns, loading buffer, wash buffer, amber collection tubes, and DEPC-treated water. The SuperScript™ Plus Direct cDNA Labeling Systems additionally include Alexa Fluor™ 555- and 647-aha-dUTP. The SuperScript™ Direct cDNA Labeling Modules contain all the same components with the exception of the purification components.

A microarray labeling kit that combines the superior performance of SuperScript III Reverse Transcriptase (RT) with the simplicity of direct labeling, plus Alexa Fluor™-aha-dUTPs

In place of an enzyme spike-in, the streamlined protocol incorporates balanced sets of novel, array-optimized, nucleotides conjugated to Alexa Fluor 555 and 647 dyes. This results in more reproducible array data with higher correlation coefficients than using standalone reagents.

In the SuperScript Plus cDNA Labeling method, anchored oligo(dT) anneals to the mRNA template. SuperScript III RT extends from the priming site, incorporating dUTP labeled with Alexa Fluor dye directly to produce labeled cDNA. Template RNA is degraded by base hydrolysis. Free nucleotides and other contaminants are removed using low-elution volume purification columns before hybridization to the microarray.

  • Optimized, all-inclusive direct labeling with balanced aha-Alexa Fluor nucleotides, enabling higher numbers of array positives
  • Improved correlation coefficients for more accurate data
  • Low elution purification columns included for ease of use

DNA and RNA Purification and Analysis, DNA Labeling, Expression Array Labeling, Gene Expression Analysis and Genotyping, Microarray Analysis, Nucleic Acid Labeling and Oligo Synthesis

TRUSTED_SUSTAINABILITY

Specifications

Product Type cDNA Labeling System
Detection Method Fluorescence
Content And Storage All SuperScript™ Direct cDNA Labeling Systems/Modules include SuperScript™ III Reverse Transcriptase, anchored Oligo(dT), random primers, control RNA, 5X first-strand buffer, 0.1 M DTT, 10 mM dNTP mix, low-elution purification columns, loading buffer, wash buffer, amber collection tubes, and DEPC-treated water. The SuperScript™ Plus Direct cDNA Labeling System/Module additionally include Alexa Fluor™ 555- and 647-aha-dUTP. The 'System' also includes purification components. See the 'Module' (L101604) if purification components are not needed.

Store purification columns, loading buffer, and wash buffer at room temperature. Store all other components at -20°C. Guaranteed stable for 6 months when properly stored.
Final Product Type cDNA (Labeled)
Label or Dye Alexa Fluor™ 555, Alexa Fluor™ 647
Includes Label or Dye Yes
Labeling Target cDNA
Labeling Method Direct Labeling
Sample Type RNA
Product Line Alexa Fluor, SuperScript
Quantity 30 Reactions
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.