Learn More
Invitrogen™ pEXP4-DEST Vector Kit
Product Code. 11580886
Change view
Click to view available options
Quantity:
6 μg
Unit Size:
6µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11580886 6 μg 6µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. Consulta la politica di reso
Codice prodotto. 11580886 Fornitore Invitrogen™ N. del fornitore V96004
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Includes
pEXP4-DEST vector, positive expression control plasmid, Tag-On-Demand slyD Mutant E. coli Extract, E. coli Reaction Buffer, Tag-On-Demand Suppressor tRNA, Methionine, DNase/RNase-free distilled water, RNase A, T7 Enzyme Mix, 2mL reaction tubes, Lumio Green Detection Kit, and the Benchmark Fluorescent Protein Standard. The pEXP4-DEST Gateway Vector Kit includes pEXP4-DEST and a control plasmid.
Enables convenient, in vitro expression of tagged or untagged protein from single vector, real-time detection of protein synthesis, easy in-gel protein detection
  • Quickly screen for expression levels of full-length protein
  • Save time and effort by cloning only once to express tagged or native protein
  • Express C-terminally tagged protein from Ultimate ORF Clone in in vitro system
  • Includes specialized E. colilysate derived from slyD mutant for cell-free synthesis: This lysate eliminates non-specific binding of LumioGreen Detection Reagent to endogenous SlyD protein, providing optimal background for recombinant protein detection
  • Clone gene of interest containing TAG (amber) stop codon into provided pEXP4-DEST
  • Features attR sites for efficient recombination, C-terminal Lumio and 6 x His tags for simplified identification and purification, and components for cell-free protein synthesis
  • In presence of ExpresswayTag-On-Demand™ Suppressor tRNA, the TAG codon is not recognized and translation continues through Lumio and 6 x His tags
  • In absence of the Suppressor tRNA, TAG stop codon is recognized and native protein is produced
  • From same vector, can rapidly determine expression level of C-terminal-tagged, full-length protein and then express native protein free from any adverse affects tag may have on its structure, activity, or molecular interactions
Cell Free Protein Expression for Functional Analysis, Cell-Free (in vitro) Expression, Cloning, Gateway Cloning, Protein Expression, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifica
Promoter T7
Product Type Gateway System Destination Expression Vector
Selection Agent (Eukaryotic) None
Content And Storage The Expressway™ Tag-On-Demand™ Cell-Free E. coli Expression System with Lumio™ Technology includes pEXP4-DEST vector (when indicated), positive expression control plasmid, Tag-On-Demand™ slyD Mutant E. coli Extract, E. coli Reaction Buffer, Tag-On-Demand™ Suppressor tRNA, Methionine, DNase/RNase-free distilled water, RNase A, T7 Enzyme Mix, 2-ml reaction tubes, Lumio™ Green Detection Kit, and the Benchmark™ Fluorescent Protein Standard. The pEXP4-DEST Gateway™ Vector Kit includes pEXP4-DEST and a control plasmid. Store vectors, detection reagent, and protein standard at -20°C. Store all other components at -80°C. Guaranteed stable for 6 months when properly stored.
Antibiotic Resistance Bacterial Ampicillin (AmpR), Zeocin™ (ZeoR)
Protein Tag Lumio
Cloning Method Gateway
Quantity 6 μg
Vector pEXP, pDEST
Product Line Expressway, Gateway
For Research Use Only. Not for use in diagnostic procedures.
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.