missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ Human DC Enrichment Kit

Product Code. 10292273
Click to view available options
Quantity:
2 x 10 mL
Unit Size:
Each
This item is not returnable. View return policy

For Research Use Only. All usage must comply with product instructions.

Product Code. 10292273

Brand: Invitrogen™ 11308D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Research Use Only. All usage must comply with product instructions.

Includes

2x10mL Depletion MyOne; Streptavidin Dynabeads and 4mL Antibody Mix containing an optimized mixture of biotinylated monoclonal antibodies towards CD3, CD14, CD16, CD19, CD56 and Glycophorin A.

Use this kit to enrich viable human dendritic cells (DC's) from PBMC by negative isolation. The kit depletes T cells, B cells, monocytes/ macrophages, granulocytes, NK cells and erythrocytes.

Use this kit to enrich viable human dendritic cells (DC's) from PBMC by negative isolation. The kit depletes T cells, B cells, monocytes/ macrophages, granulocytes, NK cells and erythrocytes. Highly enriched DC's are bead and antibody-free, and can be used in any functional assay. The cells should be further sorted to obtain high purity of the specific DC sub-populations of choice.

• Obtain high recovery (80-100%) of viable DC's of all sup-populations.
• Substantial reduction in sorting time and volume compared to sorting only, thus saving money as well.

Methodology:
An antibody mix towards the non- dendritic cells is added to the sample and allowed to bind to the cells. Dynabeads™ are added and will bind to the antibody-labeled cells during a short incubation. The bead-bound cells are quickly separated on a magnet and discarded. The remaining untouched human DC's can be directly sorted into the sup-population of choice or used in any other application.

Starting samples:
PBMC

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifica

Cell Type Dendritic Cells
Content And Storage The Kit Contains: 2 x 10 mL Depletion MyOne™ Streptavidin Dynabeads™ and 4 mL Antibody Mix containing an optimized mixture of biotinylated monoclonal antibodies towards CD3, CD14, CD16, CD19, CD56 and Glycophorin A.

Store at 2°C to 8°C.
Product Type Cell Enrichment Kit
Isolation Technology Negative Isolation
No. of Cells Processes ∼2 x 109 cells total
Output Viability >95%
Product Line DYNAL, Dynabeads
Purity or Quality Grade Research Grade
Quantity 2 x 10 mL
Sample Type PBMC
Shipping Condition Room Temperature
Starting Material Cell No. 1 x 107 PBMCs per isolation
Target Species Human
Vedi altri risultati Mostra meno risultati

For Research Use Only. Not for use in diagnostic procedures.

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato