Learn More
Intron-Binding Protein Of 160 KDa Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-23607
 View more versions of this product
Product Code. 18178721
  • 708.00 € / 0.10mg
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
Intron-Binding Protein Of 160 KDa Polyclonal antibody specifically detects Intron-Binding Protein Of 160 KDa in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Intron-Binding Protein Of 160 KDa
Polyclonal
Western Blot 1:500
O60306
AQR
Synthetic peptide corresponding to the following peptide sequence: DYILLSLVRTRAVGHLC
0.1 mg
Primary
Human, Mouse, Rat
IgG
Western Blot
Unconjugated
Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide
AQR, aquarius homolog (mouse), FSAP164, Functional Spliceosome-Associated Protein 164, IBP160, Intron-Binding Protein Aquarius
Rabbit
Affinity Purified
RUO
9716
Store at -20C. Avoid freeze-thaw cycles.