Learn More
B Braun™ Introcan Safety™ Safety Vein Indwelling Cannula
Description
- Safety venous indwelling cannula with FEP or polyurethane catheter for a venous gentle placement and uncomplicated residence time
- Handle design for one-hand puncture technique
- Excellent puncture properties due to atraumatic cannula tip with 3-facet cut geometry
- Highly transparent blood chamber
- Capillary: Four fully embedded X-ray contrast strips
- Hydrophobic blood catcher plugs
- Removable lock closure cone
- Lock approach
- Color size indicator
- Latex-free
- PVC-free
- According to ISO standard 10.555-1 / 5
- Self-activating clip technique
- Safety-Clip secures the needle tip immediately after pulling out
- Safety mechanism can not be bypassed
- Minor customization of user habits
Specifications
Specifications
| Material | FEP |
| Color | Yellow |
| Description | PVC-free |
| Needle Gauge | 24 G |
| Diameter (Metric) Inner | 0.70 mm |
| Latex Free | No |