Learn More
Hach Pacific Pty Ltd Intellical MTC101 Low Maintenance Gel Filled ORP/RedOx Electrode
Description
Intellical MTC101 is a digital, combination oxidation reduction potential (ORP/Redox) probe with a non-refillable, gel-electrolyte reference and built-in temperature sensor. MTC101 has low maintenance needs thanks to a non-refillable gel-filled single open reference junction. Provides fast, stable response in a variety of applicationsUltimate traceability in measurement historyUser alerted when re-calibration is neededCan be moved between meters without the need to re-calibrate or re-enter measurement settings
Intellical MTC101 is a digital combination oxidation reduction potential (ORP/RedOx) probe with a non-refillable, gel-electrolyte reference and built-in temperature sensor. MTC101 has low maintenance needs thanks to a non-refillable gel-filled single open reference junction. Laboratory and field models available. Intellical MTC101 provides ultimate traceability in measurement history, user alert when re-calibration is needed, ability to be moved between meters without the need to re-calibrate or re-enter measurement settings.
Standard/laboratory version:
- Suitable for use in wastewater, drinking water and aqueous media applications
- Cable length 1 or 3m
- Shockproof
- Not suitable for use with organic solvents
Rugged/field version:
- Suitable for use in external environments such as rivers, surface and ground waters, ponds, lakes, sea, drinking water reservoir for water quality, environmental and treatment process purposes
- Cable length 5, 10, 15 or 30m; Stainless steel body
- Yellow coloured cable onto which you can clip depth markers
- Waterproof up to 30 meters for 24 hours (IP68 rating)
Specifications
Specifications
| Junction | Single/Open |
| Type | Standard/Lab |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.