missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 3/CD61 Antibody (CL7319), Novus Biologicals™
Mouse Monoclonal Antibody
302.00 € - 504.00 €
Specifications
| Antigen | Integrin beta 3/CD61 |
|---|---|
| Clone | CL7319 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18639060
|
Novus Biologicals
NBP2-76484-25ul |
25 μL |
302.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18250199
|
Novus Biologicals
NBP2-76484 |
100 μL |
504.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Integrin beta 3/CD61 Monoclonal specifically detects Integrin beta 3/CD61 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Integrin beta 3/CD61 | |
| Monoclonal | |
| Purified | |
| Cancer, Cytoskeleton Markers, Phospho Specific, Signal Transduction | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
| 3690.0 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: VRDLPEELSLSFNATCLNNEVIPGLKSCMGLKIGDTVSFSIEAKVRGCPQEKEKSFTIKPVGFKDSLIVQVTFDCDCACQAQAEPNSHRCNNGNGTFECGVCRCGP | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL7319 | |
| Unconjugated | |
| Mouse | |
| Human | |
| CD61, CD61 antigen, GP3Aplatelet glycoprotein IIIa, GPIIIaintegrin beta-3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, Platelet membrane glycoprotein IIIa | |
| ITGB3 | |
| IgG1 | |
| Protein A purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title