missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 2/CD18 Antibody (MEM-48) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-379
This item is not returnable.
View return policy
Description
Integrin beta 2/CD18 Monoclonal specifically detects Integrin beta 2/CD18 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Radioimmunoassay, CyTOF-ready.
Specifications
| Integrin beta 2/CD18 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 2 ug/ml, Flow Cytometry 2 ug/ml, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:50, Immunohistochemistry-Paraffin 10 ug/ml, Immunohistochemistry-Frozen 1:200, Functional, Radioimmunoassay 1:100-1:2000, CyTOF-ready | |
| ITGB2 | |
| Leukocytes of a patient suffering from a LGL-type leukemia. | |
| Multi-step | |
| RUO | |
| Primary | |
| The antibody MEM-48 recognizes an epitope involving residues 534-546 in cysteine-rich repeat 3 of the CD18 antigen (integrin beta2 subunit; beta2 integrin). CD18 is a 90-95 kDa type I transmembrane protein expressed on all leukocytes. | |
| Store at 4C. Do not freeze. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| MEM-48 | |
| Unconjugated | |
| 959 beta subunit precursor)10, CD18 antigen, CD18beta 2, cell surface adhesion glycoprotein (LFA-1, CR3, integrin beta-2, integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1;macrophage antigen 1 (mac-1) beta subunit), integrin, beta 2 (complement component 3 receptor 3 and 4 subunit), LAD, LCAMB, leukocyte cell adhesion molecule CD18, leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C, MF17, MFI7, P150 | |
| Mouse | |
| 95 kDa | |
| 0.1 mg | |
| Apoptosis, Cytokine Research, Immunology | |
| 3689 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction