Learn More
Intedanib Piperazinyl-N4-oxide, TRC
Specifications
Specifications
| Chemical Name or Material | Intedanib Piperazinyl-N4-oxide |
| CAS | 2734666-06-7 |
| Molecular Formula | C31 H33 N5 O5 |
| Quantity | 1 mg |
| InChI Formula | InChI=1S/C31H33N5O5/c1-34(27(37)20-35-15-17-36(2,40)18-16-35)24-12-10-23(11-13-24)32-29(21-7-5-4-6-8-21)28-25-14-9-22(31(39)41-3)19-26(25)33-30(28)38/h4-14,19,32H,15-18,20H2,1-3H3,(H,33,38)/b29-28- |
| SMILES | COC(=O)c1ccc2\C(=C(\Nc3ccc(cc3)N(C)C(=O)CN4CCN(=O)(C)CC4)/c5ccccc5)\C(=O)Nc2c1 |
| IUPAC Name | methyl (3Z)-3-[[4-[methyl-[2-(4-methyl-4-oxo-1,4λ5-diazinan-1-yl)acetyl]amino]anilino]-phenylmethylidene]-2-oxo-1H-indole-6-carboxylate |
| Molecular Weight (g/mol) | 555.624 |
| Formula Weight | 555.248 |