missing translation for 'onlineSavingsMsg'
Learn More

INT-777 R-enantiomer, MedChemExpress

Product Code. 16459287
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
Unit Size:
2mg
5mg
This item is not returnable. View return policy

Product Code. 16459287

Brand: Medchem Express HY15677A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress INT-777 (R-enantiomer) is the R-enantiomer of INT-777, with EC50 of 4.79 μM for TGR5, and less potent than INT-777.

INT-777 (R-enantiomer) is the R-enantiomer of INT-777, with EC50 of 4.79 μM for TGR5, and less potent than INT-777.

Specifications

Chemical Name or Material INT-777 R-enantiomer
CAS 1198786-98-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C27H46O5
Quantity 5 mg
Synonym S-EMCA R enantiomer
Solubility Information DMSO : ≥ 100 mg/mL (221.90 mM)
SMILES O[C@@H]1CC[C@@]2(C)[C@@]([C@@H](CC)[C@@H](O)[C@]3([H])[C@]2([H])C[C@H](O)[C@@]4(C)[C@@]3([H])CC[C@@H]4[C@H](C)C[C@@H](C)C(O)=O)([H])C1
Molecular Weight (g/mol) 450.65
Formula Weight 450.65
Percent Purity 98.73%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.