missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin Antibody (IRDN/1980R), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-08836CL1
This item is not returnable.
View return policy
Description
Insulin Monoclonal antibody specifically detects Insulin in Bovine,Human,Mouse,Porcine,Rabbit samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Insulin | |
| Monoclonal | |
| CoraFluor 1 | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| Recombinant human full-length Insulin protein (IRDN) (Uniprot: P01308) | |
| 0.1 mL | |
| Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Stem Cell Markers | |
| 3630 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| IRDN/1980R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Bovine, Human, Mouse, Pig, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction