missing translation for 'onlineSavingsMsg'
Learn More

INH14, MedChemExpress

Product Code. 16451097
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16451097

Brand: Medchem Express HY114454

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress INH14 is a cell permeable inhibitor of IKKα/IKKβ, with IC50s of 8.97 and 3.59 μM, respectively. INH14 inhibits the IKKα/β-dependent TLR inflammatory response. INH14 also inhibits downstream of TAK1/TAB1 and NF-kB pathways. Anti-inflammatory and anti-cancer activity.

INH14 is a cell permeable inhibitor of IKKα/IKKβ, with IC50s of 8.97 and 3.59 μM, respectively. INH14 inhibits the IKKα/β-dependent TLR inflammatory response. INH14 also inhibits downstream of TAK1/TAB1 and NF-kB pathways. Anti-inflammatory and anti-cancer activity.

Specifications

Chemical Name or Material INH14
Physical Form Solid
CAS 200134-22-1
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C15H16N2O
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : ≥ 125 mg/mL (520.18 mM)
SMILES O=C(NC1=CC=CC=C1)NC2=CC=C(CC)C=C2
Molecular Weight (g/mol) 240.3
Formula Weight 240.3
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.