missing translation for 'onlineSavingsMsg'
Learn More

Influenza A H9N2 Neuraminidase Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

310.00 € - 507.00 €

Specifications

Antigen Influenza A H9N2 Neuraminidase
Dilution Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18660277
View Documents
Novus Biologicals
NBP3-12729-100ul
100 μg
507.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18654776
View Documents
Novus Biologicals
NBP3-12729-50ul
50 μg
310.00 €
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Influenza A H9N2 Neuraminidase Polyclonal antibody specifically detects Influenza A H9N2 Neuraminidase in Virus samples. It is validated for Western Blot, ELISA
Specifications

Specifications

Influenza A H9N2 Neuraminidase
Western Blot, ELISA
Unconjugated
Rabbit
Virus
Produced in rabbits immunized with purified, recombinant Influenza A H9N2 Neuraminidase (Accession#: NP_859038.1; His36-Ile469).
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000
Polyclonal
Purified
RUO
0.2 um filtered solution in PBS
IgG
Antigen and protein A Affinity-purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.