Learn More
Influenza A H6N1 Hemagglutinin Antibody (8A10D8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-06474-100ul
 View more versions of this product
Product Code. 18613829
  • 448.00 € / 100µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
Influenza A H6N1 Hemagglutinin Monoclonal antibody specifically detects Influenza A H6N1 Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Influenza A H6N1 Hemagglutinin
Monoclonal
Unconjugated
0.2 um filtered solution in PBS
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Influenza A virus H6N1 Hemagglutinin.
100 μg
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Western Blot, ELISA
8A10D8
Western Blot 1:500-1:1000, ELISA 1:1000-1:2000
Mouse
Protein A purified
RUO
Virus
Purified