missing translation for 'onlineSavingsMsg'
Learn More

Indole, TRC

Product Code. 30134784
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134784 25 g 25g
30005276 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134784 Supplier Toronto Research Chemicals Supplier No. TRCI57732025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-72-9
InChI Formula InChI=1S/C8H7N/c1-2-4-8-7(3-1)5-6-9-8/h1-6,9H
IUPAC Name 1H-indole
Molecular Formula C8 H7 N
Molecular Weight (g/mol) 117.15
Chemical Name or Material Indole

Specifications

Quantity 25 g
Synonym Indole, 1H-Indole, Indole (8CI), 1-Azaindene, 1-Benzazole, 2,3-Benzopyrrole, Benzo[b]pyrrole, Ketole, NSC 1964
SMILES c1ccc2[nH]ccc2c1
Formula Weight 117.0578
Percent Purity >95%
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.