Learn More
MedChemExpress Indocyanine green
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Specifications
Specifications
| Content And Storage | 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light) |
| Product Type | Dye Reagents |
| Quantity | 500 mg |
| Shipping Condition | Room temperature |
| Synonym | Foxgreen ∣ IC Green |
| CAS | 3599-32-4 |
| Molecular Weight (g/mol) | 774.96 g/mol |
| Solubility | DMSO : 33.33 mg/mL (43.01 mM; Need ultrasonic) ∣ H2O : 5 mg/mL (6.45 mM; ultrasonic and warming and heat to 60°C) |
| Target | HBV |