Learn More
Indibulin, MedChemExpress
Product Code. 16471947
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16471947
Brand: Medchem Express HY13649
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Indibulin (ZIO 301), an orally applicable inhibitor of tubulin assembly, shows potent anticancer activity with a minimal neurotoxicity. Indibulin reduces inter-kinetochoric tension, produces aberrant spindles, activates mitotic checkpoint proteins Mad2 and BubR1, and induces mitotic arrest and apoptosis.
Indibulin (ZIO 301), an orally applicable inhibitor of tubulin assembly, shows potent anticancer activity with a minimal neurotoxicity. Indibulin reduces inter-kinetochoric tension, produces aberrant spindles, activates mitotic checkpoint proteins Mad2 and BubR1, and induces mitotic arrest and apoptosis.
Specifications
Chemical Name or Material Indibulin
CAS 204205-90-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C22H16ClN3O2
Quantity 50 mg
Synonym ZIO 301 D 24851
Solubility Information DMSO : 50 mg/mL (128.26 mM; Need ultrasonic)
SMILES O=C(NC1=CC=NC=C1)C(C2=CN(CC3=CC=C(Cl)C=C3)C4=C2C=CC=C4)=O
Molecular Weight (g/mol) 389.83
Formula Weight 389.83
Percent Purity 99.47%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death