missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Indibulin

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Indibulin is a microtubule destabilizer that blocks tubulin polymerization (IC50 = 0.3 μM). Blocks cell cycle progression at metaphase and displays antitumor activity against a variety of malignancies in vitro (IC50 values ranging from 0.036 - 0.285 μM). Induces complete tumor remission in the Yoshida AH13 rat sarcoma model in vivo. Lacks neurotoxicity due to an ability to discriminate between posttranslationally modified tubulin in mature neuronal microtubules and less-modified tubulin present in immature neuronal or nonneuronal microtubules. Displays oral availability and efficacy towards MDR tumor cells.
Specifications
Specifications
| Purity | >98% |
| Quantity | 10 mg |
| Product Type | Indibulin |
| Molecular Weight (g/mol) | 389.83 |
| Inhibitors | Indibulin |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction