missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Indacaterol maleate

Product Code. p-7214505 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 16534434

Brand: Thermo Scientific Chemicals 464652500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 753498-25-8
Molecular Formula C28H32N2O7
Molecular Weight (g/mol) 508.57
MDL Number MFCD20526769,MFCD18782702
InChI Key IREJFXIHXRZFER-UHFFFAOYNA-N
IUPAC Name 5-{2-[(5,6-diethyl-2,3-dihydro-1H-inden-2-yl)amino]-1-hydroxyethyl}-8-hydroxy-1,2-dihydroquinolin-2-one; but-2-enedioic acid
SMILES OC(=O)C=CC(O)=O.CCC1=C(CC)C=C2CC(CC2=C1)NCC(O)C1=C2C=CC(=O)NC2=C(O)C=C1

Specifications

Melting Point 195.0°C to 197.0°C
Quantity 250 mg
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in dmso
Formula Weight 508.57
Chemical Name or Material Indacaterol maleate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.