Learn More
Ecolab™ Incidin™ OxyWipe S
Product Code. 15951312
Change view
Click to view available options
Packaging Quantity:
600 Wipes
Unit Size:
Pack of 600
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity Unit Size
15951312 600 Wipes Pack of 600
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15951312 Supplier Ecolab™ Supplier No. 3082240
  • ONLINE EXCLUSIVE PRICE
    338.10 €
    249.88 € / Pack of 600
    Save 88.22 €
    26% Off
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
Sporicidal ready-to-use cleaner and disinfectant with Hi-Speed H2O2 a patented solution with broad spectrum efficacy
  • Hi-Speed H2O2™
  • Active ingredient decomposes into water and oxygen 
  • Sporicidal & virucidal wipe for all surfaces
  • Excellent material compatibility
Specifications
Packaging Quantity 600 Wipes