missing translation for 'onlineSavingsMsg'
Learn More

Inavolisib, MedChemExpress

Product Code. p-200099328
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16496137

Brand: Medchem Express HY101562

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GDC-0077 (RG6114) is a potent, orally available, and selective PI3Kα inhibitor (IC50=0.038 nM). GDC-0077 (RG6114) exerts its activity by binding to the ATP binding site of PI3K, thereby inhibiting the phosphorylation of PIP2 to PIP3. GDC-0077 (RG6114) is more selective for mutant versus wild-type PI3Kα.

GDC-0077 (RG6114) is a potent, orally available, and selective PI3Kα inhibitor (IC50=0.038 nM). GDC-0077 (RG6114) exerts its activity by binding to the ATP binding site of PI3K, thereby inhibiting the phosphorylation of PIP2 to PIP3. GDC-0077 (RG6114) is more selective for mutant versus wild-type PI3Kα.

Specifications

Chemical Name or Material Inavolisib
CAS 2060571-02-8
Color Tan
Purity Grade Notes Research
Molecular Formula C18H19F2N5O4
Quantity 5 mg
Synonym GDC-0077 RG6114
Solubility Information DMSO : 110 mg/mL (270.02 mM; Need ultrasonic)
SMILES C[C@@H](C(N)=O)NC1=CC=C(C2=NC(N3[C@H](C(F)F)COC3=O)=CN2CCO4)C4=C1
Molecular Weight (g/mol) 407.37
Formula Weight 407.37
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.