missing translation for 'onlineSavingsMsg'
Learn More

Inarigivir soproxil, MedChemExpress

Product Code. 16486977
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16486977

Brand: Medchem Express HY109035

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Inarigivir soproxil (SB9200) is an agonist of innate immunity and shows potent antiviral activity against resistant HCV variants, with EC50s of 2.2 and 1.0 μM for HCV 1a/1b in cells of genotype 1 HCV replicon systems. Inarigivir soproxil, an orally bioavailable prodrug of SB 9000, has broad-spectrum antiviral activity against RNA viruses including HCV, norovirus, respiratory syncytial virus and influenza and HBV.

Inarigivir soproxil (SB9200) is an agonist of innate immunity and shows potent antiviral activity against resistant HCV variants, with EC50s of 2.2 and 1.0 μM for HCV 1a/1b in cells of genotype 1 HCV replicon systems. Inarigivir soproxil, an orally bioavailable prodrug of SB 9000, has broad-spectrum antiviral activity against RNA viruses including HCV, norovirus, respiratory syncytial virus and influenza and HBV.

Specifications

Chemical Name or Material Inarigivir soproxil
Physical Form Solid
CAS 942123-43-5
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C25H34N7O13PS
For Use With (Application) COVID-19-anti-virus
Synonym SB9200 GS-9992
Solubility Information DMSO : 80 mg/mL (113.70 mM; Need ultrasonic) ∣H2O : 10 mg/mL (14.21 mM; ultrasonic and warming and heat to 60°C)
SMILES CO[C@H]1[C@H](N(C=CC2=O)C(N2)=O)O[C@H](CO)[C@H]1OP(OC[C@H]3O[C@@H](N4C5=NC=NC(N)=C5N=C4)C[C@@H]3O)(SCOC(OC(C)C)=O)=O
Molecular Weight (g/mol) 703.62
Formula Weight 703.62
Percent Purity 98.08%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.