missing translation for 'onlineSavingsMsg'
Learn More

Inarigivir, MedChemExpress

Product Code. 16426397
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16426397

Brand: Medchem Express HY101954

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Inarigivir (ORI-9020) is a dinucleotide antiviral drug that can significantly reduce liver HBV DNA in transgenic mice expressing hepatitis B virus. Inarigivir (ORI-9020) act as a RIG-I agonist to activate cellular innate immune responses.

Inarigivir (ORI-9020) is a dinucleotide antiviral drug that can significantly reduce liver HBV DNA in transgenic mice expressing hepatitis B virus. Inarigivir (ORI-9020) act as a RIG-I agonist to activate cellular innate immune responses.

Specifications

Chemical Name or Material Inarigivir
Physical Form Solid
CAS 475650-36-3
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C20H26N7O10PS
For Use With (Application) COVID-19-anti-virus
Synonym ORI-9020 SB-9000
Solubility Information DMSO : 200 mg/mL (340.43 mM; Need ultrasonic)
SMILES NC1=C(N=CN2[C@@H]3O[C@H](COP(O[C@H]4[C@@H](OC)[C@H](N5C=CC(NC5=O)=O)O[C@@H]4CO)(S)=O)[C@@H](O)C3)C2=NC=N1
Molecular Weight (g/mol) 587.5
Formula Weight 587.5
Percent Purity 98.82%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.