missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B Braun™ IN-Stopper Sealing Cones
Click to view available options
Unit Size:
Pack of 100
Description
- Smooth, hygienic, contamination-free outer surface
- Contact-free Luer taper
- Priming volume of 0.16 mL
- Latex-free
- Easy and convenient in handling, can also be used as closing cones for female accesses
- Yellow in color
- Provided in packs of 100
Specifications
Specifications
| Color | Yellow |
| For Use With (Application) | Intermittent injections through injection membrane |
| Latex Free | Yes |
| Product Type | Stopper/Sealing Cone |
| Quantity | 100/Pk. |
| Volume (Metric) | 0.16 mL (Priming) |
| Connection Type | Luer Taper |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction