missing translation for 'onlineSavingsMsg'
Learn More

IMR-1, MedChemExpress

Product Code. p-200098429
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16423018 10 mg 10mg
16414326 100 mg 100mg
16409586 50 mg 50mg
16405757 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16423018 Supplier Medchem Express Supplier No. HY100431

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress IMR-1 is a novel class of Notch inhibitor targeting the transcriptional activation with an IC50 of 26 μM. IMR-1 prevents the recruitment of Mastermind-like 1 (Maml1) to the Notch Ternary Complex (NTC) on chromatin, inhibits Notch target gene transcription and dramatically inhibits tumor growth.

IMR-1 is a novel class of Notch inhibitor targeting the transcriptional activation with an IC50 of 26 μM. IMR-1 prevents the recruitment of Mastermind-like 1 (Maml1) to the Notch Ternary Complex (NTC) on chromatin, inhibits Notch target gene transcription and dramatically inhibits tumor growth.

Specifications

Chemical Name or Material IMR-1
CAS 310456-65-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H15NO5S2
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (282.96 mM)
SMILES O=C(OCC)COC1=CC=C(/C=C(SC(N2)=S)\C2=O)C=C1OC
Molecular Weight (g/mol) 353.41
Formula Weight 353.41
Percent Purity 98.88%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.