Learn More
IMR-1, MedChemExpress
Product Code. 16423018
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16423018
Brand: Medchem Express HY100431
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress IMR-1 is a novel class of Notch inhibitor targeting the transcriptional activation with an IC50 of 26 μM. IMR-1 prevents the recruitment of Mastermind-like 1 (Maml1) to the Notch Ternary Complex (NTC) on chromatin, inhibits Notch target gene transcription and dramatically inhibits tumor growth.
IMR-1 is a novel class of Notch inhibitor targeting the transcriptional activation with an IC50 of 26 μM. IMR-1 prevents the recruitment of Mastermind-like 1 (Maml1) to the Notch Ternary Complex (NTC) on chromatin, inhibits Notch target gene transcription and dramatically inhibits tumor growth.
Specifications
Chemical Name or Material IMR-1
CAS 310456-65-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H15NO5S2
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (282.96 mM)
SMILES O=C(OCC)COC1=CC=C(/C=C(SC(N2)=S)\C2=O)C=C1OC
Molecular Weight (g/mol) 353.41
Formula Weight 353.41
Percent Purity 98.88%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death