Learn More
IMP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16205-1-AP-20UL
 View more versions of this product
Product Code. 16895744
  • 145.00 € / 20µL
Please to purchase this item. Need a web account? Register with us today!
Description
Description
IMP4 forms a ternary complex with IMP3.
Specifications
Specifications
IMP4
Polyclonal
Unconjugated
IMP4
BXDC4, IMP4, U3 snoRNP protein IMP4
Rabbit
Antigen Affinity Chromatography
RUO
27993, 316317, 92856
-20°C
Liquid
Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5PQR5, Q8VHZ7, Q96G21
Imp4
IMP4 Fusion Protein Ag9175
20 μL
Primary
Rat, Human, Mouse