Learn More
MP Biomedicals™ Immuno™ Albumin, Bovine, Low Endotoxin Powder (Heat-Shock)
Used to solubilize lipids, also used as blocking agent in Western blots or ELISA applications.
Brand: MP Biomedicals™ 08810683
Additional Details : CAS : 9048-46-8 Weight : 0.10000kg
Description
- Derived from bovine blood collected at USDA licensed facility Prepared by heat-shocked fractionate method
Specifications
ELISA, Immunoassay, Mammalian Cell Culture, Western Blot | |
Immuno™ Albumin, Bovine, Low Endotoxin (Heat Shock) | |
100g | |
0° to 5°C |
∽66 | |
7.1 | |
Bovine Blood | |
Powder |
For Research Use Only.