Learn More
MP Biomedicals™ Immuno™ Albumin, Bovine, Low Endotoxin Powder (Heat-Shock)
Used to solubilize lipids, also used as blocking agent in Western blots or ELISA applications.
Brand:  MP Biomedicals™ 08810683
Additional Details : CAS : 9048-46-8 Weight : 0.10000kg
Product Code. 11474226
  • 973.08 € / 100g
Estimated Shipment: 26-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description
Description
  • Derived from bovine blood collected at USDA licensed facility
  • Prepared by heat-shocked fractionate method
Specifications
Specifications
ELISA, Immunoassay, Mammalian Cell Culture, Western Blot
Immuno™ Albumin, Bovine, Low Endotoxin (Heat Shock)
100g
0° to 5°C
∽66
7.1
Bovine Blood
Powder