Learn More
Testo™ Immersion/Penetration Probe
Product Code. 10120651
Click to view available options
Temperature Range (Metric):
-200°C to +1000°C
Unit Size:
Each
Product Code. 10120651
Brand: Testo™ 0602 5792
Only null left
Add to basket
Add to basket
Specifications
Type Immersion/Penetration Probe
Certifications/Compliance EN 60584-2
Response Time 5 sec.
Temperature Range (Metric) -200°C to +1000°C