Learn More
Zeiss Immersion oil Immersol 518 F / 23°C, oiler 20ml
Brand: Zeiss 433802-9010-000
Immersion oil Immersol 518 F / 23°C, oiler 20ml
Safety and Handling
- Immersion oil Immersol 518 F / 23°C, oiler 20ml
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
Supplemental information
- MIXTURE LIST-Contain: Adipic acid-di(8-methyl-tricyclo (5.2.1.0.2.6.) decane) ester, benzyl benzoate
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.