Learn More
Imirestat, MedChemExpress
Product Code. 16497008
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497008 10 mg 10mg
16491599 1 mg 1mg
16449557 5 mg 5mg
16420576 20 mg 20mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16497008 Supplier Medchem Express Supplier No. HY16255
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Imirestat (AL 1576) is an aldose reductase inhibitor, used for the treatment of diabetes.
Imirestat (AL 1576) is an aldose reductase inhibitor, used for the treatment of diabetes.
Specifications
Chemical Name or Material Imirestat
CAS 89391-50-4
Color White
Purity Grade Notes Research
Molecular Formula C15H8F2N2O2
Quantity 10 mg
Synonym AL 1576 Alcon 1576 HOE 843
Solubility Information DMSO : 83.33 mg/mL (291.13 mM; Need ultrasonic)
SMILES O=C1NC2(C(N1)=O)C3=C(C4=C2C=C(F)C=C4)C=CC(F)=C3
Molecular Weight (g/mol) 286.23
Formula Weight 286.23
Percent Purity 98.42%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism