missing translation for 'onlineSavingsMsg'
Learn More

Imiquimod, 99%

Product Code. p-7042853
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15416139 100 mg 100mg
15406139 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15416139 Supplier Thermo Scientific Alfa Aesar Supplier No. J63990.MC
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99011-02-6 | C14H16N4 | 240.31 g/mol

Imiquimod is a caspase 3 activator which acts as an immunomodulator and displays antiviral and anti-tumor activity. It is a patient-applied cream used for the treatment of genital warts and basal cell carcinoma. It is also used to cure actinic keratosis on the face and scalp. It belongs to a group of drugs called immune response modifiers, which work by activating the immune system to fight abnormal skin growth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dimethyl sulfoxide and dimethyl formamide. Slightly soluble in water. Insoluble in ethanol.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99011-02-6
Molecular Formula C14H16N4
Molecular Weight (g/mol) 240.31
MDL Number MFCD00866946
InChI Key DOUYETYNHWVLEO-UHFFFAOYSA-N
Synonym imiquimod, aldara, zyclara, beselna, 1-2-methylpropyl-1h-imidazo 4,5-c quinolin-4-amine, zartra, imiquimod acetate, 1-isobutyl-1h-imidazo 4,5-c quinolin-4-amine, 4-amino-1-isobutyl-1h-imidazo 4,5-c quinoline
PubChem CID 57469
ChEBI CHEBI:36704
IUPAC Name 1-(2-methylpropyl)imidazo[4,5-c]quinolin-4-amine
SMILES CC(C)CN1C=NC2=C1C1=CC=CC=C1N=C2N

Specifications

Melting Point 292°C to 294°C
Color White
Quantity 100 mg
UN Number UN2811
Merck Index 144922
Solubility Information Soluble in dimethyl sulfoxide and dimethyl formamide; Slightly soluble in water; Insoluble in ethanol.
Formula Weight 240.31
Percent Purity 99%
Physical Form Solid
Chemical Name or Material Imiquimod

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.