missing translation for 'onlineSavingsMsg'
Learn More

Imidazole-d3, TRC

Product Code. 30397355
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30397355 50 mg
30401727 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30397355 Supplier Toronto Research Chemicals Supplier No. TRCI35020350MG
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6745-43-3
Molecular Formula C3HD3N2
Molecular Weight (g/mol) 71.1
Synonym 1H-Imidazole-2,4,5-d3 (9CI), Imidazole-2,4,5-d3 (6CI, 7CI, 8CI), Imidazole-d3, Imidazole-D3
SMILES [2H]C1=C(N=C(N1)[2H])[2H]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C3H4N2/c1-2-5-3-4-1/h1-3H,(H,4,5)/i1D,2D,3D
Formula Weight 71.0563
Chemical Name or Material Imidazole-d3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.