Learn More
Imidazole-d3, TRC
Product Code. 30401727
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30401727 500 mg
30397355 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30401727 Supplier Toronto Research Chemicals Supplier No. TRCI350203500MG
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6745-43-3
Molecular Formula C3HD3N2
Molecular Weight (g/mol) 71.1
Synonym 1H-Imidazole-2,4,5-d3 (9CI), Imidazole-2,4,5-d3 (6CI, 7CI, 8CI), Imidazole-d3, Imidazole-D3
SMILES [2H]C1=C(N=C(N1)[2H])[2H]
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C3H4N2/c1-2-5-3-4-1/h1-3H,(H,4,5)/i1D,2D,3D
Formula Weight 71.0563
Chemical Name or Material Imidazole-d3