Learn More
Velp Scientifica™ Imhoff Cone
Velp Scientifica™ Imhoff Cone for use with FCS and JLT Series Flocculators.
Brand: Velp Scientifica™ A00001002
64.60 EUR valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Additional Details : Weight : 0.24000kg
Description
Velp Scientifica™ Imhoff Cone for use with FCS and JLT Series Flocculators.
- Transparent for visability
- Available in glass or plastic
- Available in graduated
- For use with Flocculators
Specifications
1000 mL | |
FCS, JLT | |
Imhoff Cone | |
Plastic |