missing translation for 'onlineSavingsMsg'
Learn More

Imatinib Mesylate, MedChemExpress

Product Code. 16431336
Encompass_Preferred
Click to view available options
Quantity:
200 mg
500 mg
1 g
5 g
Unit Size:
1g
200mg
500mg
5g
This item is not returnable. View return policy

Product Code. 16431336

Brand: Medchem Express HY50946

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Imatinib Mesylate (STI571 Mesylate) is a tyrosine kinases inhibitor that inhibits c-Kit, Bcr-Abl, and PDGFR (IC50=100 nM) tyrosine kinases.

Imatinib Mesylate (STI571 Mesylate) is a tyrosine kinases inhibitor that inhibits c-Kit, Bcr-Abl, and PDGFR (IC50=100 nM) tyrosine kinases.

Specifications

Chemical Name or Material Imatinib Mesylate
CAS 220127-57-1
Color White
Purity Grade Notes Research
Molecular Formula C30H35N7O4S
Quantity 200 mg
Synonym STI571 MesylateCGP-57148B Mesylate
Solubility Information DMSO : 125 mg/mL (211.97 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (84.79 mM)
SMILES O=S(O)(C)=O.O=C(NC1=CC=C(C(NC2=NC=CC(C3=CC=CN=C3)=N2)=C1)C)C4=CC=C(CN5CCN(CC5)C)C=C4
Molecular Weight (g/mol) 589.71
Formula Weight 589.71
Percent Purity 99.43%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.