Learn More
ILT1/CD85h/LILRA2 Mouse anti-Human, Alexa Fluor 647, Clone: 600007, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB6364R-100UG
Description
ILT1/CD85h/LILRA2 Monoclonal specifically detects ILT1/CD85h/LILRA2 in Human samples. It is validated for Flow Cytometry.Specifications
| ILT1/CD85h/LILRA2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q8N149 | |
| LILRA2 | |
| Mouse myeloma cell line NS0-derived recombinant human LILRA2/CD85h/ILT1 Pro17-Ser437 Accession # Q8N149 | |
| RUO | |
| 11027 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 600007 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD85 antigen-like family member H, CD85h, CD85h antigen, ILT1, ILT-1, ILT1CD85H, Leukocyte immunoglobulin-like receptor 7, leukocyte immunoglobulin-like receptor subfamily A member 2, leukocyte immunoglobulin-like receptor subfamily A member 2 soluble, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 2, LIR7, LIR7LIR-7Immunoglobulin-like transcript 1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human LILRA2/CD85h/ILT1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) ILT2, 3, 4, 5, rhLIR6, or rhLIR8 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only (RUO)