missing translation for 'onlineSavingsMsg'
Learn More

Ilginatinib maleate, MedChemExpress

Product Code. 16447828
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16447828 10 mg 10mg
16490288 5 mg 5mg
16483777 50 mg 50mg
16427656 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16447828 Supplier Medchem Express Supplier No. HY19631
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Ilginatinib maleate (NS-018 maleate) is a highly active and orally bioavailable JAK2 inhibitor, with an IC50 of 0.72 nM, 46-, 54-, and 31-fold selectivity for JAK2 over JAK1 (IC50, 33 nM), JAK3 (IC50, 39 nM), and Tyk2 (IC50, 22 nM).

Ilginatinib maleate (NS-018 maleate) is a highly active and orally bioavailable JAK2 inhibitor, with an IC50 of 0.72 nM, 46-, 54-, and 31-fold selectivity for JAK2 over JAK1 (IC50, 33 nM), JAK3 (IC50, 39 nM), and Tyk2 (IC50, 22 nM).

Specifications

Chemical Name or Material Ilginatinib maleate
CAS 1354799-87-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C25H24FN7O4
Quantity 10 mg
Synonym NS-018 maleate
Solubility Information DMSO : ≥ 30 mg/mL (59.35 mM) ∣H2O : 1 mg/mL (1.98 mM; Need ultrasonic)
SMILES CN1N=CC(C2=CC(N[C@H](C3=CC=C(F)C=C3)C)=NC(NC4=NC=CN=C4)=C2)=C1.O=C(O)/C=C\C(O)=O
Molecular Weight (g/mol) 505.5
Formula Weight 505.5
Percent Purity 97.04%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.