missing translation for 'onlineSavingsMsg'
Learn More

IL36 Beta/IL1F8 Rabbit anti-Human, Polyclonal, Proteintech

Product Code. 16838084 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16838084 20 μL 20µL
16828084 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16838084 Supplier Proteintech Supplier No. 180431AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IL-36B is is a member of the interleukin 1 cytokine family whose gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. IL-36B is thought to activate the NF-kappaB pathway through IL-1 receptor family members IL-1RL2 and IL-1RAcP. Like the related proteins IL-36A and IL-36G, IL-36B requires post-translational processing for full agonist activity, but the cleavage mechanism is currently unknown. The IL-36 cytokines have been suggested to amplify Th1 responses by enhancing proliferation and Th1 polarization of naive CD4+ T cells.

Specifications

Antigen IL36 Beta/IL1F8
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene IL36B
Gene Accession No. Q9NZH7
Gene Alias FIL1, FIL1 (ETA), FIL1 eta, FIL1H, IL 1 eta, IL 1F8, IL 1H2, IL1 ETA, IL1F8, IL1H2, Interleukin 1 eta, Interleukin 1 family member 8, Interleukin 1 homolog 2
Gene Symbols IL36B
Host Species Rabbit
Immunogen IL36 Beta/IL1F8 Fusion Protein Ag12675
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27177
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.