missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress IL-6R alpha Protein, Human (CHO)
Specifications
Specifications
| Content And Storage | Stored at -20°C. After reconstitution, it is stable at 4°C for 1 week or -20°C for longer. It is recommended to freeze aliquots at -20°C or -80°C for extended storage. |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Quantity | 10 μg |
| Shipping Condition | Room temperature |
| Molecular Weight (g/mol) | 54-56 kDa |
| Synonym | rHuIL-6R∣ IL-6RA∣ IL-6R 1∣ CD126∣ Membrane glycoprotein 80 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?