missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress IL-6 Protein, Human

Product Code. 16459728
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16459728 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16459728 Supplier MedChemExpress Supplier No. HYP7044

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress IL-6 Protein, Human is a potent inducer of the synthesis of acute phase proteins in adult human hepatocytes.

Specifications

Content And Storage Stored at -20°C. After reconstitution, it is stable at 4°C for 1 week or -20°C for longer. It is recommended to freeze aliquots at -20°C or -80°C for extended storage.
Quantity 10 μg
Shipping Condition Room temperature
Molecular Weight (g/mol) Approximately 21,000 Da
Synonym rHuIL-6∣ BSF-2∣ CDF∣ Hybridoma growth factor∣ IFN-beta-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.