missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-6 Human Magnetic Kit for Luminex™ Platform

Product Code. 15461164
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15461164 100 Tests 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15461164 Supplier Invitrogen™ Supplier No. LHC0061M

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

IL-6 Human Magnetic Kit for Luminex™ Platform

The Human IL-6 Magnetic Bead Kit for the Luminex™ platform is specifically designed for quantification of low levels of human IL-6 in serum, plasma, and tissue culture supernatant samples. By mixing this kit with other targets from our human magnetic kit portfolio, one can obtain more data from each sample, saving both money and time compared to more traditional methods (such as ELISA). This kit is recommended to be run with the Human Cytokine Magnetic Buffer Reagent Kit (Cat. No. LHB0001M). This assay employs magnetic beads, facilitating automation, decreasing hands-on time, and increasing throughput and precision. The assay is suitable for use with the Luminex™ 200, FLEXMAP 3D™, and MAGPIX™ systems.
  • Superior performance—accurate, reproducible, and sensitive quantitation of multiple proteins
  • High quality—fully qualified antibodies produce excellent specificity and sensitivity
  • Fast and easy protocols—perform your assays and analyze your data typically in less than one day
Luminex xMAP technology—effective and proven analytical tool

The Human IL-6 Magnetic Bead Kit is based on xMAP™ technology. The use of a suspension bead-based technology enables the multiplexing capabilities of the Luminex assays. Magnetic microspheres are internally dyed with red and infrared fluorophores of differing intensities. Each bead has a unique number, or bead region, allowing differentiation of one bead from another. Beads covalently bound to different antibodies can be mixed in the same assay. Upon completion of the sandwich immunoassay, magnetic beads must be measured using a Luminex™ detection system (MAGPIX™, 100™/200™, or FLEXMAP 3D™). The instrument uses xPONENT™ software to distinguish bead color (analyte) and R-PE fluorescence intensity (assay signal strength) to quantify target(s).

Magnetic beads let you do more with your time

Magnetic bead-based assays enable automation, decrease hands-on time, and increase throughput and precision. Using the fast and easy Luminex protocols, you can obtain results in as little as 3.5 hours. This line of assays was carefully designed and tested for maximum sensitivity, range, and correlation. Using the same components as our polystyrene bead-based assays allows for performance with identical quality and consistency. Standard curves generated with this assay system extend over several orders of magnitude of reagent concentration, while the sensitivity and quality of the assays are comparable to ELISAs and our polystyrene bead-based assays, enabling superior performance.

More validation for confidence in your results

The Human IL-6 Magnetic Bead Kit has been validated using the same rigorous criteria that distinguishes Luminex assays from other commercially available assays. Our fully qualified antibodies help ensure the specificity and sensitivity of our assays. Every singleplex kit and premixed multiplex kit comes with a product insert that outlines the specifications for each marker in the assay.

TRUSTED_SUSTAINABILITY

Specifications

Description IL-6 Human Magnetic Kit, 100 Tests, IL6 Gene Symbol, 3569 Gene ID, R-PE Conjugate, Magnetic Bead Type, Human Species, Cytokines and Receptors Protein Family, Use with Luminex™ Instruments, Ideal for Protein Assays and Analysis
Product Type ELISA Kit
Quantity 100 Tests
Species Human
Detection Method Fluorescence
Conjugate R-PE
Bead Type Magnetic
For Use With (Application) Protein biology, luminex assays and other bead-based assays, protein assays and analysis
For Use With (Equipment) Luminex Instruments
Gene ID (Entrez) 3569
Gene Symbol IL6
Substrate Type Cytokines & Receptors
Kit Contents Antibody-coated capture beads, standard, detector antibody and lot-specific technical data sheet
Content And Storage 2°C to 8°C
Regulatory Status For research use only (RUO)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.