missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-5R alpha/CD125 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody has been used in 1 publication
Brand: R&D Systems AF553-SP
This item is not returnable.
View return policy
Description
IL-5R alpha/CD125 Polyclonal specifically detects IL-5R alpha/CD125 in Mouse samples. It is validated for Western Blot.Specifications
| IL-5R alpha/CD125 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD125, CD125 antigen, CDw125CD125, HSIL5R3, IL-5 receptor subunit alpha, IL5R, IL5R alpha, IL-5R subunit alpha, IL5RA, IL-5RA, IL-5R-alpha, interleukin 5 receptor type 3, interleukin 5 receptor, alpha, interleukin-5 receptor alpha chain, interleukin-5 receptor alpha subunit, interleukin-5 receptor subunit alpha, MGC26560 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3568 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| P21183 | |
| IL5RA | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-5 R alpha/CD125 Asp18-His339 Accession # P21183 | |
| 25 μg | |
| Primary | |
| Detects mouse IL-5 R alpha/CD125 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) IL-5 R alpha is observed and less than 1% cross-reactivity with recombinant mouse (rm) IL-4 R, rhIL-9 R, rhIL-13 R alpha 1, and rhIL-13 R alpha 2 is observed. | |
| Mouse | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?