Learn More
IL-17RD/SEF Antibody (400210), PE/Atto594, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB2276PEATT594
Product Code. 30032858
  • 674.00 € / 0.10mL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
IL-17RD/SEF Monoclonal antibody specifically detects IL-17RD/SEF in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-17RD/SEF
Monoclonal
PE/ATTO 594
PBS
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
54756
Store at 4C in the dark. Do not freeze.
IgG2a
Flow Cytometry
400210
Flow Cytometry
FLJ35755, IL-17RDIL-17 receptor D, IL17Rhom, IL17RLMDKFZp434N1928, interleukin 17 receptor D, interleukin-17 receptor D, Interleukin-17 receptor-like protein, MGC133309, Sef homolog, SEFhSef
Mouse myeloma cell line NS0-derived recombinant mouse IL-17 RD/SEF, Gly28-Arg299, Accession # Q8JZL1
0.1 mL
Primary
Mouse
Purified