missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-13R alpha 2 Antibody (018), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-89441CL1
This item is not returnable.
View return policy
Description
IL-13R alpha 2 Monoclonal antibody specifically detects IL-13R alpha 2 in Human samples. It is validated for Flow Cytometry
Specifications
| IL-13R alpha 2 | |
| Monoclonal | |
| CoraFluor 1 | |
| cancer/testis antigen 19, CD213a2, CD213a2 antigen, CT19, IL-13 receptor subunit alpha-2, IL13BP, IL13R, IL-13R, IL-13R subunit alpha-2, IL-13RA2, IL-13R-alpha-2, interleukin 13 binding protein, interleukin 13 receptor alpha 2 chain, interleukin 13 receptor, alpha 2, interleukin-13 receptor subunit alpha-2, Interleukin-13-binding protein | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-13R alpha 2 (Accession#: NP_000631.1; Met1-Leu342). | |
| 0.1 mL | |
| Cancer | |
| 3598 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Flow Cytometry | |
| 018 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction