Learn More
IKA™ R1827 Plate Stand
Product Code. 10545861 Shop All IKA Products
Click to view available options
Diameter (Metric) Shaft:
16 mm
Unit Size:
Each
Product Code. 10545861
Brand: IKA™ 0003160200
Please to purchase this item. Need a web account? Register with us today!
With slip resistant foil.
  • Support rod diameter of 16mm
  • Maximum load of 5kg
  • 200mm W x 316mm D x 800mm H
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Shaft 16 mm
Height (Metric) 1000 mm
Quantity 1 Each
Type Plate Stand
Width (Metric) 200 mm