Learn More
Proteintech IHCeasy NCBP2 Ready-To-Use IHC Kit
Product Code. 18030546 Shop All Proteintech Products
Click to view available options
Quantity:
50 Tests
Unit Size:
50 tests
Product Code. 18030546
Brand: Proteintech KHC162150T
Please to purchase this item. Need a web account? Register with us today!
KHC1621 is a ready-to-use IHC kit for staining of NCBP2.
KHC1621 is a ready-to-use IHC kit for staining of NCBP2. The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifications
Quantity 50 Tests
Form Liquid