Learn More
Abnova™ IGH/CCND1 DY Translocation FISH Probe
Specifications
Specifications
| Content And Storage | Store at 4°C in the dark |
| Format | Tube |
| For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
| Gene | immunoglobulin heavy locus |
| Gene Alias | IGH,IGHDY1,MGC72071,MGC88774 |
| Gene Symbol | IGH |
| Gene ID (Entrez) | 3492;595 |
| Product Type | DNA/RNA |
| Quantity | 200 μL |
| Form | Liquid |